Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD84 Monoclonal Antibody (CD84.1.21), PE
GREENER_CHOICE

Catalog No. PIMA528609
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528609 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528609 Supplier Invitrogen™ Supplier No. MA528609
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope of CD84, a single chain cell surface glycoprotein of 64-82 kDa, predominantly expressed B cells, monocytes, platelets and some T cells. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.

CD84 is expressed on mature B cells and on B cell lines, including pre B cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD84
Applications Flow Cytometry
Classification Monoclonal
Clone CD84.1.21
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene CD84
Gene Accession No. Q9UIB8
Gene Alias A130013D22Rik; CD84; CD84 antigen; CD84 antigen (leukocyte antigen); CD84 molecule; CDw84; cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; leukocyte differentiation antigen CD84; LY9B; mCD84; RGD1564553; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5
Gene Symbols CD84
Host Species Mouse
Immunogen CD84-transfected 3.19 cell line.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8832
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.