Learn More
Invitrogen™ CD84 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592970
Description
Immunogen sequence: KDSEIFTVNG ILGESVTFPV NIQEPRQVKI IAWTSKTSVA YVTPGDSETA PVVTVTHRNY YERIHALGPN YNLVISDLRM EDAGDYKADI NTQADPYTTT KRYNLQIYRR LGKPKITQSL MASVNSTCNV TLTCSVEKEE KNVTYNWSPL GEEGNVLQIF QTPEDQELTY TCTAQNPVSN NSDSISARQL CADIAMGFRT HHTG; Positive Samples: Jurkat, THP-1; Cellular Location: Cell membrane, Single-pass type I membrane protein.
CD84 is expressed on mature B cells and on B cell lines, including pre B cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.
Specifications
CD84 | |
Polyclonal | |
Unconjugated | |
CD84 | |
A130013D22Rik; CD84; CD84 antigen; CD84 antigen (leukocyte antigen); CD84 molecule; CDw84; cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; leukocyte differentiation antigen CD84; LY9B; mCD84; RGD1564553; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8832 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UIB8 | |
CD84 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-225 of human CD84 (NP_0038651). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.