Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD84/SLAMF5 Antibody (004), Biotin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289266B
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for ELISA.Specifications
CD84/SLAMF5 | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
8832 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
4 | |
ELISA | |
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD84/SLAMF5 (NP_003865.1; Met1-Gly225). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction