Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD84/SLAMF5 Antibody (152-1D5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311538AF350
Description
CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD84/SLAMF5 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
152-1D5 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
Spleen cells of a patient with hairy cell leukemia | |
0.1 mL | |
CD Markers, Immunology, Stem Cells | |
8832 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction