Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD84/SLAMF5 Antibody (6), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289267AF532
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, ELISA.Specifications
CD84/SLAMF5 | |
Monoclonal | |
Alexa Fluor 532 | |
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD84/SLAMF5 extracellular domain (Met 1-Gly 225; NP_003865.1). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
6 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
8832 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction