Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD84/SLAMF5 Antibody (CD84.1.21), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065929R
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry.Specifications
CD84/SLAMF5 | |
Monoclonal | |
DyLight 550 | |
50 mM sodium borate with 0.05% sodium azide | |
CD84 | |
Mouse pre-B cell line 300.19 transfected with CD84 cDNA. | |
Primary | |
NB100-65929 recognizes the human CD84 cell surface antigen, a highly glycosylated cell surface antigen of 64-82kD. CD84 is predominantly expressed by B lymphocytes, monocytes and macrophages, although lower levels of expression have also been reported on T cells, granulocytes and platelets. | |
Store at 4°C in the dark. |
Flow Cytometry | |
CD84.1.21 | |
Flow Cytometry | |
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
Mouse | |
0.1 mL | |
8832 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction