Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CD84/Slamf5, Rat, Unlabeled, Clone: 1D3,

Catalog No. MABF929MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF929MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF929MI Supplier MilliporeSigma™ Supplier No. MABF929
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SLAM family member 5 (UniProt Q18PI6; also known as Leukocyte differentiation antigen CD84, Signaling lymphocytic activation molecule 5, CD84) is encoded by the Cd84 (also known as Slamf5) gene (Gene ID 12523) in murine species. CD84 belongs to the 7-member (SLAM/CD150, CD48/SLAMF2, Ly-9/CD229, 2B4/CD244, CD84/SLAMF5, Ly108/NTB-A/CD352, CRACC) signaling lymphocytic activation molecule family (SLAMF) of receptors broadly expressed in hematopoietic cells. 2B4 and CD48 interact with eath other, while rest of the family members act as self-ligands, where two identical molecules on the surface of two different cells can interact as receptor and ligand to each other. SLAMF members carry one or more copies of immunoreceptor tyrosine-based switch motif or ITSM (TxYxxV/I) in their cytoplasmic tails. Upon phosphorylation on the tyrosine residue, ITSM mediates the recruitment of SH2 domain-containing signaling molecules such as SLAM-associated protein (SAP), including SAP in T-, NK, and NK T-cells, EAT-2A and EAT-2B (murine) in NK cells and APCs. Murine CD84 is produced with a signal peptide sequence (a.a. 1-21), the removal of which yields the mature CD84 with an extracellular region (a.a. 22-221) that contains a V-type Ig-like domain (a.a. 26-129) and a C2-type Ig-like domain (a.a. 132-206), followed by a single transmembrane (a.a. 222-242) and a cytoplasmic tail (a.a. 243-329) with two copies of ITSM (a.a. 263-268 and 298-303). Alternative splicings produce three CD84 isoforms, with spliced isoform 2 produced as a truncated extracellular domain with an alternative C-terminal sequence (from 128-RLKTPKITQSLIS-140 to 128-KLWQHGALDLLLI-140) and no transmembrane or cytoplasmic sequence (a.a. 141-329 missing).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD84/Slamf5
Applications Flow Cytometry
Classification Monoclonal
Clone 1D3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Cd84, Slamf5
Gene Accession No. Q18PI6
Host Species Rat
Immunogen Human IgG1 Fc-tagged recombinant mouse CD84 extracellular domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_038517.1
Test Specificity Clone 1D3 immunostained the surface of murine CD84-transfected, but not untranfected, BI-141 murine T-cells (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305). Reactivity toward spliced isoform 2 has not been determined.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.