Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD85d (ILT4) Monoclonal Antibody (27D6), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. 5014554
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-145-54 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-145-54 Supplier Invitrogen™ Supplier No. 16514881
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody 27D6 recognizes the CD85d protein also known as ILT4, LIR-2. It is a member of the ILT (immunoglobulin-like transcript)/ LIR (leuokocyte Ig-like receptor) MIR (monocytes Ig-like receptor) family. CD85d is a 110 kDa single transmembrane receptor containing 3 Ig-like domains and a long cytoplasmic domain containing ITIMs (immunoreceptor tyrosine-based inhibitory motifs). Expression is confined to monocytes, macrophages and dendritic cells. The ligand for CD85d is non-classical class I HLA-G, -B and G1. Upon ligand binding, tyrosine phosphatase SHP-1 interacts with the ITIM. Interaction of HLA-G with ILT4/CD85d is thought to contribute to tolerance during fetal implantation. Additionally it is the levels of ILT3 and ILT4 on DCs that suggests a role in organ allograft rejection. The monoclonal antibody 27D6 has been shown to block binding of ligand (HLA-G tetramer). Applications Reported: This 27D6 antibody has been reported for use in vitro in blocking assays and in flow cytometric analysis. Applications Tested: This 27D6 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated fo...

CD85d, also known as LILRB2 or ILT4, is a member of the immunoglobulin-like transcript (ILT), leukocyte Ig-like receptor (LIR), and monocyte Ig-like receptor (MIR) family. It is a 110 kDa single transmembrane receptor with three Ig-like domains and a long cytoplasmic domain containing immunoreceptor tyrosine-based inhibitory motifs (ITIMs). CD85d is expressed on monocytes, macrophages, and dendritic cells. The receptor binds to non-classical class I molecules such as HLA-G, -B, and G1. Upon ligand binding, the tyrosine phosphatase SHP-1 interacts with the ITIMs, transducing a negative signal that inhibits immune response stimulation. This interaction is thought to contribute to tolerance during fetal implantation and plays a role in controlling inflammatory responses and cytotoxicity, helping to focus the immune response and limit autoreactivity. CD85d is also involved in organ allograft rejection, with the levels of ILT3 and ILT4 on dendritic cells suggesting its role in this process. The LILRB2 gene, located in a gene cluster at chromosomal region 19q13.4, encodes multiple transcript variants with different isoforms, highlighting its complexity and importance in immune regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85d (ILT4)
Applications Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
Clone 27D6
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene LILRB2
Gene Accession No. Q8N423
Gene Alias CD85 antigen-like family member D; CD85D; CD85d antigen; Ig-like transcript 4; ILT4; ILT-4; leucocyte Ig-like receptor B2; leukocyte immunoglobulin like receptor B2; leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRB2; Lilrb3; LIR2; LIR-2; LIR2CD85D; MIR10; MIR-10; MIR10LILRA6; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10; Neutrophil immunoglobulin-like receptor 1; Nilr1; NILR-1; paired-Ig-like receptor B; Pirb; type 1 one-pass transmembrane Ig-like receptor
Gene Symbols LILRB2
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10288
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.