Learn More
Invitrogen™ CD85g (ILT7) Monoclonal Antibody (eBio17G10.2 (17G10.2)), PE, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 12517942

Description
Description: The eBio17G10.2 monoclonal antibody reacts with human immunoglobulin-like transcript 7 (ILT7), also known as CD85g. ILT7 is part of the leukocyte immunoglobulin-like receptor (LIR) gene family and associates with Fc epsilon R1 gamma to form a functional complex which transduces ITAM-mediated signals that negatively modulates TLR-induced type I IFN production by human plasmacytoid dendritic cells. ILT7 is expressed exclusively on plasmacytoid dendritic cells and is absent in monocyte derived DCs. Treatment with IL-3 or CpG causes a decrease in expression. Double staining of human PBMCs with eBio17G10.2 and anti-pDC marker BDCA2 yields virtually complete co-staining. Cross linking of ILT7 with eBio17G10.2 antibody causes reduced transcription of IFN alpha1, IFN alpha2, IFN alpha4 and IFN beta. Applications Reported: This eBio17G10.2 (17G10.2) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio17G10.2 (17G10.2) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
ILT7 is a pDC-specific IgG-like transcript and is a member of the leukocyte immunoglobulin-like receptor gene family. This receptor, in complex with FcER1a, transduces ITAM-mediated signals which negatively modulate type I IFN production by pDC. ILT7 is thus a potential target to regulate Type 1 IFN production by pDC, which play a major role during viral infection, auto-immune diseases and anti-cancer immunity. Additionally, ILT7, along with CD123 and BDCA-2, can be used to identify or classify subsets of pDCalong with TLR expression and TLR mediated immune responses.
Specifications
CD85g (ILT7) | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P59901 | |
LILRA4 | |
Affinity chromatography | |
RUO | |
23547 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBio17G10.2 (17G10.2) | |
PE | |
LILRA4 | |
CD85 antigen-like family member G; CD85g; ILT7; ILT-7; Immunoglobulin-like transcript 7; leucocyte Ig-like receptor A4; leukocyte immunoglobulin like receptor A4; leukocyte immunoglobulin-like receptor subfamily A member 4; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 4; LILRA4 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.