Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD85g (ILT7) Mouse, anti-Human, R718, Clone: 17G10.2, BD OptiBuild™

Catalog No. BDB752195 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752195 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752195 Supplier BD Biosciences Supplier No. 752195
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 17G10.2 monoclonal antibody specifically binds to human Immunoglobulin Like Transcript 7 (ILT7), also known as CD85g. The ILT7 protein is encoded by the LILRA4 (Leukocyte Immunoglobulin Like Receptor subfamily A member 4) gene. ILT7 is a member of immunoglobulin-like transcripts (ILT) or leukocyte immunoglobulin-like receptor (LIR) gene family. ILT7 is selectively expressed on plasmacytoid dendrtic cells (pDC) but not on myeloid dendritic cells or other peripheral blood leukocytes. In response to IL-3, IL-3 receptor complex signaling downregulates ILT7 expression by pDC. ILT7 associates with the FcεRIγ signal adapter protein to form a receptor complex that can signal through activation of an immunoreceptor-based tyrosine activation motif (ITAM)-mediated signaling pathway. Signaling through the ILT7-FcεRIγ receptor complex negatively regulates the innate immune functions of pDC. ILT7 crosslinking inhibits TLR responses by pDC such as the stimulated production of type I interferon and other cytokines. Bone marrow stromal cell antigen 2 (BST2) has been identified as a biological ligand for ILT7.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85g (ILT7)
Applications Flow Cytometry
Classification Monoclonal
Clone 17G10.2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.