Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561961 Supplier Invitrogen™ Supplier No. PA561961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DHRLSWTLNS HQHNHGKFQA LFPMGPLTFS NRGTFRCYGY ENNTPYVWSE PSD Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.

ILT7 is a pDC-specific IgG-like transcript and is a member of the leukocyte immunoglobulin-like receptor gene family. This receptor, in complex with FcER1a, transduces ITAM-mediated signals which negatively modulate type I IFN production by pDC. ILT7 is thus a potential target to regulate Type 1 IFN production by pDC, which play a major role during viral infection, auto-immune diseases and anti-cancer immunity. Additionally, ILT7, along with CD123 and BDCA-2, can be used to identify or classify subsets of pDCalong with TLR expression and TLR mediated immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LILRA4
Gene Accession No. P59901
Gene Alias CD85 antigen-like family member G; CD85g; ILT7; ILT-7; Immunoglobulin-like transcript 7; leucocyte Ig-like receptor A4; leukocyte immunoglobulin like receptor A4; leukocyte immunoglobulin-like receptor subfamily A member 4; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 4; LILRA4
Gene Symbols LILRA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CD85g (ILT7). Recombinant protein control fragment (Product #RP-99158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.