Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD85h (ILT1) Monoclonal Antibody (135.4), PE, eBioscience™

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 12511942

Encompass_Preferred

Catalog No. 50-108-02


Only null left
Add to Cart

Description

Description

Description: The monoclonal antibody 135.4 recognizes ILT1, also known as CD85h, LIR7 and LILRA2. ILT1 is a member of the large family of ILT molecules (Immunologulin-like Transcripts) also known as Leukocyte Ig-like Receptors (LIRs) and Monocyte/Macrophage Ig-like Receptors (MIRs). ILT1 contains a short cytoplasmic domain. Through the cytoplasmic domain ILT1 associates with the Fc receptor gamma chain but not DAP12 resulting in a stimulatory signal. ILT1 may act as a receptor for MHC Class I antigens. Expression of ILT1 is mainly found on monocytes, macrophages, DC and weakly on granulocytes. ILT1 is thought to play a role in the TH1/Th2 balance as well as TLR signaling. The monoclonal antibody 135.4 has been reported to have activating properties upon crosslinking which result in calcium influx. Applications Reported: This 135.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 135.4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD85h (ILT1)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q8N149
LILRA2
Affinity Chromatography
RUO
11027
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
135.4
PE
LILRA2
CD85 antigen-like family member H; CD85h; ILT1; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; leukocyte immunoglobulin like receptor A2; leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LILRA2; LIR7; LIR-7
Rat
100 Tests
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.