Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD85j (ILT2) Monoclonal Antibody (HP-F1), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD85j (ILT2) |
---|---|
Clone | HP-F1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H043T03Y04A
![]() |
Invitrogen™
H043T03Y04A |
100 Tests |
Each for $430.00
|
|
|||||
H043T02Y04A
![]() |
Invitrogen™
H043T02Y04A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD85j (ILT2) Monoclonal antibody specifically detects CD85j (ILT2) in Human samples. It is validated for Flow CytometrySpecifications
CD85j (ILT2) | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
Q8NHL6 | |
10859 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HP-F1 | |
Flow Cytometry | |
NovaFluor Yellow 660 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B | |
IgG1 κ | |
Affinity chromatography | |
LILRB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title