Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD85j (ILT2) Mouse anti-Human, BUV615, Clone: GHI/75, BD OptiBuild™

Catalog No. BDB752329 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752329 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752329 Supplier BD Biosciences Supplier No. 752329
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD85j (ILT2) Monoclonal antibody specifically detects CD85j (ILT2) in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85j (ILT2)
Applications Flow Cytometry
Classification Monoclonal
Clone GHI/75
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu CD85j BUV615 GHI/75 50 μg
Host Species Mouse
Immunogen Human Hairy Cell Leukemia Spleen Cells
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD85j (ILT2)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2b κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.