Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD85k (Gp49b) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596569 Supplier Invitrogen™ Supplier No. PA596569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAGPLPKPTL WAEPGSVISW GNSVTIWCQG TLEAREYRLD KEESPAPWDR QNPLEPKNKA RFSIPSMTED YAGRYRCYYR SPVGWSQPSD PLELVMTGAY SKPTLSALPS PLVTSGKSVT LLCQSRSPMD TFLLIKERAA HPLLHLRSEH GAQQHQAEFP MSPVTSVHGG TYRCFSSHGF SHYLLSHPSD PLELIVSGSL EGPRPSPTRS VSTAAGPEDQ PLMPTGSVPH SGLRRHWE; Positive Samples: HL-60, Mouse liver, Mouse heart; Cellular Location: Cell membrane, Single-pass type I membrane protein.

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85k (Gp49b)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene LILRB4
Gene Accession No. Q64281, Q8NHJ6
Gene Alias CD85 antigen-like family member K; CD85K; glycoprotein 49 B; glycoprotein 49b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; mast cell surface glycoprotein Gp49B; Monocyte inhibitory receptor HM18
Gene Symbols LILRB4, Lilrb4a
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11006, 14728, 292594
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.