Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD85k Mouse anti-Human, Brilliant Violet 650, Clone: ZM3.8, BD Optibuild™

Catalog No. BDB742808 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742808 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742808 Supplier BD Biosciences Supplier No. 742808
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ZM3.8 monoclonal antibody specifically binds to CD85k which is also known as Immunoglobulin-like transcript 3 (ILT-3) and Monocyte inhibitory receptor HM18. CD85k is encoded by LILRB4 (Leukocyte immunoglobulin-like receptor, subfamily B member 4) and is expressed as a type I transmembrane glycoprotein. CD85k is comprised of two C2-type Ig domains, a transmembrane region, and a cytoplasmic tail with three immunoreceptor tyrosine-based inhibitory motifs (ITIMs). CD85k is selectively expressed by myeloid antigen presenting cells including monocytes, macrophages, and dendritic cells. CD85k functions as an inhibitory receptor. ILT3 can also function in antigen capture and presentation. It is efficiently internalized upon cross-linking, and delivers its ligand to an intracellular compartment where it is processed and ultimately presented by APCs to T cells. Thus, ILT3 is a novel inhibitory receptor that can negatively regulate activation of APCs and can be used by APCs for antigen uptake. ILT3 expression is upregulated on tolerogenic dendritic cells that can induce regulatory T cells.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85k
Classification Monoclonal
Clone ZM3.8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias LILRB4, ILT-3, HM18, LIRB4, LILRB5, LIR-5
Host Species Mouse
Immunogen Human ILT3 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.