Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD86 (B7-2) Mouse anti-Human, Rhesus, Cynomolgus, Baboon, R718, Clone: 2331 (FUN-1), BD Horizon™
Mouse Monoclonal Antibody
$132.00 - $402.00
Specifications
Antigen | CD86 (B7-2) |
---|---|
Clone | 2331 (FUN-1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
CD86 (B7-2) Monoclonal antibody specifically detects CD86 (B7-2) in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow CytometrySpecifications
CD86 (B7-2) | |
Flow Cytometry | |
R718 | |
Mouse | |
Human, Rhesus Macaque, Cynomolgus, Baboon | |
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
2331 (FUN-1) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HBL-1 Cell Line | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title