Learn More
Invitrogen™ CD86 Monoclonal Antibody (1B3)

Description
MA5-15697 targets CD86 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15697 immunogen is purified recombinant fragment of human CD86 expressed in E. Coli. MA5-15697 detects CD86 which has a predicted molecular weight of approximately 52kDa.
Specifications
Specifications
| Antigen | CD86 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1B3 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | CD86 |
| Gene Accession No. | P42081, P42082 |
| Gene Alias | activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.