Learn More
Invitrogen™ CD86 Monoclonal Antibody (GL1), PE-Cyanine7

Description
CD86 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD86, CD86 (B7-2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | GL1 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE-Cyanine7 |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.1 |
| Gene | CD86 |
| Gene Accession No. | P42082 |
| Gene Alias | activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.