Learn More
CD86 Mouse anti-Human, FITC, Clone: K86, Abnova™
Mouse monoclonal antibody raised against human CD86.
Supplier: Abnova Corporation MAB12416
Description
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined. [provided by RefSeq]
Specifications
CD86 | |
Monoclonal | |
FITC | |
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
B7-2/B70/CD28LG2/LAB72/MGC34413 | |
Mouse | |
Protein A/G affinity chromatography | |
RUO | |
942 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
K86 | |
Mouse monoclonal antibody raised against human CD86. | |
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
CD86 | |
CD86 | |
50 Reactions | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.