Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: C86/2160R, Abnova™

Catalog No. 89339685 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-685 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-685 Supplier Abnova Corporation Supplier No. RAB00670
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD86 Monoclonal antibody specifically detects CD86 in Human, Mouse, Rat samples. It is validated for ELISA, WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen CD86
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone C86/2160R
Conjugate Unconjugated
Description CD86 molecule
Gene CD86
Gene Alias B7-2, B70, CD28LG2, LAB72, MGC34413
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CD86
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.