Learn More
CD87 Mouse anti-Human, Brilliant Violet 786, Clone: VIM5 (also known as VIM-5), BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743100
Description
The VIM5 monoclonal antibody specifically recognizes CD87. CD87 is also known as the urokinase plasminogen activator receptor (uPA-R). CD87 is a 50-65 kDa single chain glycosylphosphatidylinositol (GPI)-linked glycoprotein. uPA-R was clustered as CD87 in the Fifth Human Leukocyte Differentiation Antigens Workshop. It is expressed on neutrophils, monocytes, macrophages, dendritic cells, endothelial cells, activated T cells, fibroblasts, and smooth muscle. CD87 plays a role in cell-associated fibrinolytic activity and tissue invasion. This antibody may be useful in the study of myeloid cell differentiation.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
CD87 | |
VIM5 (also known as VIM-5) | |
Brilliant Violet 786 | |
uPA-R, U-PAR, PLAUR, URKR | |
Human THP-1 Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.