Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD87 Mouse anti-Human, BUV661, Clone: VIM5 (also known as VIM-5), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 750465
Description
CD87 Monoclonal antibody specifically detects CD87 in Human samples. It is validated for Flow Cytometry
Specifications
CD87 | |
Monoclonal | |
Brilliant Ultraviolet 661 | |
uPA-R; U-PAR; PLAUR; URKR | |
Human THP-1 Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
VIM5 (also known as VIM-5) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction