Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD87 Mouse, anti-Human, R718, Clone: VIM5, BD OptiBuild™

Catalog No. BDB751966 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751966 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751966 Supplier BD Biosciences Supplier No. 751966
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The VIM5 monoclonal antibody specifically recognizes CD87. CD87 is also known as the urokinase plasminogen activator receptor (uPA-R). CD87 is a 50-65 kDa single chain glycosylphosphatidylinositol (GPI)-linked glycoprotein. uPA-R was clustered as CD87 in the Fifth Human Leukocyte Differentiation Antigens Workshop. It is expressed on neutrophils, monocytes, macrophages, dendritic cells, endothelial cells, activated T cells, fibroblasts, and smooth muscle. CD87 plays a role in cell-associated fibrinolytic activity and tissue invasion. This antibody may be useful in the study of myeloid cell differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD87
Applications Flow Cytometry
Classification Monoclonal
Clone VIM5
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.