Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD88 Mouse, anti-Mouse, R718, Clone: 20/70, BD OptiBuild™

Catalog No. BDB752094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752094 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752094 Supplier BD Biosciences Supplier No. 752094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 20/70 monoclonal antibody specifically binds to CD88, which is also known as, Complement component 5a receptor 1 (C5ar1), C5a anaphylatoxin chemotactic receptor, or C5a receptor (C5aR). CD88 belongs to the rhodopsin family of seven-transmembrane G protein-coupled receptors and mediates its effects by binding to the complement activation product, C5a. CD88 is expressed on leucocytes including, neutrophils, monocytes, macrophages, and eosinophils. CD88 may also be expressed on alveolar epithelial cells, endothelial cells, neural stem cells, oligodendrocytes, and on parenchymal cells of lung, liver, kidney and heart. Binding of the proinflammatory C5a anaphylatoxin to CD88 results in G-protein coupling and intracellular signal transduction that mediates intracellular calcium release, chemotaxis, degranulation, cytokine production, and superoxide anion production. The 20/70 antibody can block binding of C5a to CD88 and thus inhibit its biological functions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD88
Applications Flow Cytometry
Classification Monoclonal
Clone 20/70
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.