Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD88 Rat anti-Mouse, Brilliant Violet 711, Clone: 20/70, BD Optibuild™

Catalog No. BDB743773 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743773 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743773 Supplier BD Biosciences Supplier No. 743773
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 20/70 monoclonal antibody specifically binds to CD88, which is also known as, Complement component 5a receptor 1 (C5ar1), C5a anaphylatoxin chemotactic receptor, or C5a receptor (C5aR). CD88 belongs to the rhodopsin family of seven-transmembrane G protein-coupled receptors and mediates its effects by binding to the complement activation product, C5a. CD88 is expressed on leucocytes including, neutrophils, monocytes, macrophages, and eosinophils. CD88 may also be expressed on alveolar epithelial cells, endothelial cells, neural stem cells, oligodendrocytes, and on parenchymal cells of lung, liver, kidney and heart. Binding of the proinflammatory C5a anaphylatoxin to CD88 results in G-protein coupling and intracellular signal transduction that mediates intracellular calcium release, chemotaxis, degranulation, cytokine production, and superoxide anion production. The 20/70 antibody can block binding of C5a to CD88 and thus inhibit its biological functions.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD88
Classification Monoclonal
Clone 20/70
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd88, C5ar1, C5a-R, C5aR, C5r1, C5a Ligand, C5a anaphylatoxin receptor
Host Species Rat
Immunogen Mouse C5aR Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.