Learn More
Description
The 20/70 monoclonal antibody specifically binds to CD88, which is also known as, Complement component 5a receptor 1 (C5ar1), C5a anaphylatoxin chemotactic receptor, or C5a receptor (C5aR). CD88 belongs to the rhodopsin family of seven-transmembrane G protein-coupled receptors and mediates its effects by binding to the complement activation product, C5a. CD88 is expressed on leucocytes including, neutrophils, monocytes, macrophages, and eosinophils. CD88 may also be expressed on alveolar epithelial cells, endothelial cells, neural stem cells, oligodendrocytes, and on parenchymal cells of lung, liver, kidney and heart. Binding of the proinflammatory C5a anaphylatoxin to CD88 results in G-protein coupling and intracellular signal transduction that mediates intracellular calcium release, chemotaxis, degranulation, cytokine production, and superoxide anion production. The 20/70 antibody can block binding of C5a to CD88 and thus inhibit its biological functions.
Specifications
Specifications
| Antigen | CD88 |
| Classification | Monoclonal |
| Clone | 20/70 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Cd88, C5ar1, C5a-R, C5aR, C5r1, C5a Ligand, C5a anaphylatoxin receptor |
| Host Species | Rat |
| Immunogen | Mouse C5aR Transfected Cell Line |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.