Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD89 Monoclonal Antibody (MIP8a), PE, Invitrogen™
Mouse Monoclonal Antibody
Specifications
| Antigen | CD89 |
|---|---|
| Clone | MIP8a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
This antibody recognizes the human CD89 cell surface antigen, a 50-75 kDa cell surface glycoprotein that is also known as the IgA receptor (Fc alpha R). CD89 is expressed by peripheral blood monocytes and neutrophils. MIP8a blocks binding of IgA to the Fc alpha R, and also inhibits neutrophil phagocytosis of IgA complexes. Reconstitute with 1 mL distilled water.
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| CD89 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| 2204 | |
| Primary | |
| Antibody |
| MIP8a | |
| Monoclonal | |
| Lyophilized | |
| RUO | |
| P24071 | |
| IgG1 | |
| 4° C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title