Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD89 Mouse, anti-Human, R718, Clone: A59, BD OptiBuild™

Catalog No. BDB752119 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752119 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752119 Supplier BD Biosciences Supplier No. 752119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A59 monoclonal antibody specifically recognizes CD89. CD89 is the Fc receptor for IgA (FcαR), a 55-75 kDa glycoprotein expressed exclusively on cells of granulocytic and monocyte/macrophage lineages in peripheral blood, but not on lymphocytes. It is also expressed on promyelocytes in bone marrow and in the cytoplasm of fixed monocytes. This mAb does not block IgA binding and is more efficient than native IgA ligands in the isolation of FcαR molecules. FcαR plays a role in triggering phagocytosis and respiratory burst.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD89
Applications Flow Cytometry
Classification Monoclonal
Clone A59
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.