Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8A, Rabbit anti-Human, FITC, Clone: N21-G, Abnova™

Catalog No. 89998671 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-671 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-671 Supplier Abnova Corporation Supplier No. MAB16006
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen CD8A
Applications Flow Cytometry
Classification Monoclonal
Clone N21-G
Conjugate FITC
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS (10 mg/mL BSA, 0.05% Sodium Azide).
Gene CD8a molecule
Gene Alias CD8/Leu2/MAL/p32
Gene Symbols CD8A
Host Species Rabbit
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.