Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8A Rat anti-Mouse, APC-Cy7, Clone: 53-6.7, Abnova™
Rat monoclonal antibody raised against Cd8a.
Supplier: Abnova Corporation MAB12215
Specifications
| CD8A | |
| Monoclonal | |
| APC-Cyanine7 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| BB154331/Ly-2/Ly-35/Ly-B/Lyt-2 | |
| Rat | |
| Affinity Chromatography | |
| RUO | |
| 12525 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry | |
| 53-6.7 | |
| Rat monoclonal antibody raised against Cd8a. | |
| In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
| Cd8a | |
| Cd8a | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction