Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8a, Rat Anti-Mouse, RB613, 53-6.7, BD Horizon™
Mouse Monoclonal Antibody
$138.00 - $346.00
Specifications
Antigen | CD8a |
---|---|
Clone | 53-6.7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD8a Monoclonal antibody specifically detects CD8a in Mouse (QC Testing) samples. It is validated for Flow CytometrySpecifications
CD8a | |
0.2 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Cd8a; CD8 alpha chain; Ly-2; Lyt2; Lyt-2; Ly-35; Ly-B | |
Mouse Spleen Cells or Thymocyte Membranes | |
Primary |
53-6.7 | |
Flow Cytometry | |
RB613 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12525 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title