Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (YTS 105.18), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252659JF549
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
CD8 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD8A | |
Ly-2-transfected mouse L cells. | |
0.1 mL | |
Primary | |
Recognizes a non polymorphic epitope on the mouse CD8 alpha chain. This antibody has been reported to block MHC I dependent T cell responses in vitro and in vivo. | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
YTS 105.18 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence | |
CD8, CD8a molecule, LEU2, p32 | |
Rabbit | |
Protein A or G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Signal Transduction, Stem Cell Markers | |
925 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction