Learn More
CD8b Monoclonal Antibody (eBio341 (341)), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$3335.00 - $9810.00
Specifications
Antigen | CD8b |
---|---|
Clone | eBio341 (341) |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-008-038
![]() |
LIFE TECHNOLOGIES
16008038 |
5 mg |
Each for $3,335.00
|
|
|||||
16-008-0025
![]() |
LIFE TECHNOLOGIES
160080025 |
25 mg |
Each for $9,810.00
|
|
|||||
Description
The eBio341 MAb reacts with rat CD8beta. CD8beta is a member of the immunoglobulin superfamily which heterodimerizes with CD8alpha to form the CD8 antigen expressed on CD4+CD8+, double-positive thymocytes, the majority of MHC class I-restricted peripheral T cells, and on some macrophages. CD8beta expression is absent in athymic mice, which are populated by T-cells expressing CD8alpha homodimers. This has led to the idea that CD8beta expression is dependent on the development of T cells in the thymus. CD8 binds to MHC class I expressed on the surface of antigen-presenting cells during antigen presentation, and participates in T-cell receptor signal transduction through association with the kinase Lck.
The CD8B antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified.Specifications
CD8b | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
24931 | |
Primary | |
4°C | |
Cd8b |
eBio341 (341) | |
Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) | |
Functional Grade | |
Mouse | |
Rat | |
P05541 | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.