Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8b Monoclonal Antibody (SIDI8BEE), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $440.00
Specifications
Antigen | CD8b |
---|---|
Clone | SIDI8BEE |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H057T03R06A
![]() |
Invitrogen™
H057T03R06A |
100 Tests |
Each for $440.00
|
|
|||||
H057T02R06A
![]() |
Invitrogen™
H057T02R06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD8b Monoclonal antibody specifically detects CD8b in Human, Non-human Primate samples. It is validated for Flow CytometrySpecifications
CD8b | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human, Monkey | |
P10966 | |
926 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
SIDI8BEE | |
Flow Cytometry | |
NovaFluor Red 755 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
IgG1 κ | |
Affinity chromatography | |
Cd8b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title