Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8b Mouse Anti-Rat, BUV395, Clone: 341, BD Optibuild™

Catalog No. BDB742921 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742921 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742921 Supplier BD Biosciences Supplier No. 742921
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 341 monoclonal antibody specifically recognizes theβchain of the CD8 antigen on most thymocytes and a subpopulation of mature T lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). The CD8αandβchains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and thymus-dependent T suppressor/cytotoxic cells, whereas intestinal intraepithelial lymphocytes, many CD8+ T cells of athymic rats, many activated CD4+ T cells, and most NK cells express CD8a without CD8b. It has been suggested that the expression of the CD8a/CD8b heterodimer is restricted to thymus-derived T lymphocytes. CD8 is an antigen co-receptor on the T cell surface which interacts with MHC class I molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Macrophages have also been reported to express CD8αandβchains, which are involved in signal transduction. The 341 mAb blocks proliferative and cytotoxic in vitro responses of CD8+ effectors to allogeneic cells.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8b
Classification Monoclonal
Clone 341
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd8b, CD8beta, CD8 beta, CD8-beta, CD8β
Host Species Mouse
Immunogen CD8-positive Wistar rat splenic T-cell hybridomas
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.