Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8b Rat anti-Mouse, BV605, Clone: YTS156.7.7.rMAb (also known as YTS156.7.7; YTS156), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 755236
This item is not returnable.
View return policy
Description
CD8b Monoclonal antibody specifically detects CD8b in Mouse samples. It is validated for Flow cytometry (Qualified)
Specifications
| CD8b | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Chromatography | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| YTS156.7.7.rMAb (also known as YTS156.7.7; YTS156) | |
| Brilliant Violet 605 | |
| Cd8b1; Cd8b; Ly-3; Lyt-3; lymphocyte antigen 3 | |
| Mouse Thymocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction