Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (IVA50) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500494
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF.Specifications
CD9 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 μg/mL, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:50, CyTOF-ready | |
CD9 | |
Bovine thrombocytes. | |
Protein A purified | |
RUO | |
Primary | |
The antibody IVA50 reacts with CD9 antigen (bovine), a 24 kDa single transmembrane polypeptide expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes. | |
Store at 4C. Do not freeze. | |
IgG2 |
Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF | |
IVA50 | |
Unconjugated | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
24 kDa | |
0.1 mg | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928 | |
Human, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction