Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (IVA50), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500494F
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine samples. It is validated for Flow Cytometry.Specifications
CD9 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CD9 | |
Bovine thrombocytes. | |
0.1 mL | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928 | |
Human, Bovine | |
Purified |
Flow Cytometry | |
IVA50 | |
Flow Cytometry | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody IVA50 reacts with CD9 antigen (bovine), a 24 kDa single transmembrane polypeptide expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes. | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction