Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (MM0180-5H15), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212184AF488
Description
CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for Flow Cytometry.Specifications
CD9 | |
Monoclonal | |
Alexa Fluor 488 | |
P21926 | |
CD9 | |
Human recombinant CD9 | |
0.1 mL | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Flow Cytometry | |
MM0180-5H15 | |
Western Blot, Flow Cytometry | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction