Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (P1/33/2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250154AF405
Description
CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD9 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD9 | |
Acute lymphoblastic leukaemia cells | |
0.1 mL | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
P1/33/2 | |
Flow Cytometry, Immunohistochemistry | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction