Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (P1/33/2), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250154PEATT594
Description
CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for Flow Cytometry.Specifications
CD9 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
CD9 | |
Acute lymphoblastic leukaemia cells | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
P1/33/2 | |
Flow Cytometry | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
Protein A purified | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction