Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Antibody (RH1A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260966
Description
CD9 Monoclonal specifically detects CD9 in Bovine, Canine, Feline, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD9 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Mouse | |
Unpurified | |
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
928 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
RH1A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD9 | |
Mononuclear leukocytes from: rat and horse | |
0.1 mg | |
Primary | |
Bovine, Canine, Feline, Rabbit | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction