Learn More
Invitrogen™ CD9 Monoclonal Antibody (MM2/57)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA180307
Description
Predicted to react with mustelid based on sequence homology. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD9 antibody, clone MM2/57 recognizes human leukocyte antigen MIC3 also known as MRP-1 or CD9.
CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.
Specifications
CD9 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P21926, P30932, P40239, Q8WMQ3 | |
CD9 | |
Human platelet membrane. | |
100 μg | |
Primary | |
Human, Canine, Rabbit, Bovine, Pig, Feline, Horse, Mink, Rhesus Macaque | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
MM2/57 | |
Unconjugated | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Mouse | |
Protein G | |
RUO | |
100059512, 100141395, 100345844, 280746, 397067, 493874, 611695, 712262, 928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.