Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1077S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1077S 1 mL Concentrated
MS1077S0 100 μL Concentrated
MS1077S1 500 μL Concentrated
MS1077R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1077S Supplier Epredia™ Supplier No. MS1077S

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody.

CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. It belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre-B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41/CD61 (GPIIb/GPIIIa).

Host Species: Mouse

Clone: 72F6

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding the major extracellular loop of the human CD9

Molecular Weight: 24-27kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD9/Motility-Related Protein-1 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 72F6
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the major extracellular loop of the human CD9
Purification Method Tonsil
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.