Learn More
Epredia™ Lab Vision™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 72F6
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding the major extracellular loop of the human CD9
Molecular Weight: 24-27kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD9/Motility-Related Protein-1 Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 72F6 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding the major extracellular loop of the human CD9 |
| Purification Method | Tonsil |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.