Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, Clone: HI9a, Abnova™

Catalog No. 89933891 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-891 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-891 Supplier Abnova Corporation Supplier No. MAB12535
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human CD9.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It can modulate cell adhesion and migration and also trigger platelet activation and aggregation. In addition, the protein appears to promote muscle cell fusion and support myotube maintenance. [provided by RefSeq]

Specifications

Antigen CD9
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone HI9a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CD9.
Dilution Flow Cytometry (0.5 ug/106 cells) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias 5H9/BA2/BTCC-1/DRAP-27/GIG2/MIC3/MRP-1/P24/TSPAN29
Gene Symbols CD9
Host Species Mouse
Immunogen Human CD9
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.