Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD9 Mouse anti-Human, Hamster, Canine, Feline, Monkey, Rabbit, Raccoon, FITC, Clone: MM2/57, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 931002
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD9 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
MM2/57 | |
FITC | |
Mouse | |
RUO | |
Canine, Feline, Hamster, Human, Monkey, Rabbit, Raccoon | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction