Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595868
Description
Immunogen sequence: SHKDEVIKEV QEFYKDTYNK LKTKDEPQRE TLKAIHYALN CCGLAGGVEQ FISDICPKKD VLETFTVKSC PDAIKEVFDN KFHI; Positive Samples: Mouse kidney; Cellular Location: Cell membrane, Membrane, Multi-pass membrane protein.
CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.
Specifications
CD9 | |
Polyclonal | |
Unconjugated | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12527, 24936, 928 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7.19 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P21926, P40240, P40241 | |
CD9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 112-195 of human CD9 (NP_0017601). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction