Learn More
CD9 Rat anti-Mouse, BUV496, Clone: KMC8 (also known as KMC 8), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | KMC8 (also known as KMC 8) |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Cd9; Tetraspanin-29; Tspan29 |
| Host Species | Rat |
| Immunogen | (C57BL/6 x DBA/2)F1 mouse bone marrow-derived stromal cell line BMS2 |
| Quantity | 50 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.